Learn More
Selleck Chemical LLC Bromoenol lactone-E1878-100MG
SDP
Catalog No. 503137796
Encompass_Preferred
Bromoenol lactone((6E)-Bromoenol lactone) is a selective irreversible potent inhibitor of calcium-independent phospholipase A2 (iPLA2 ) with an IC50 value of 7 M It prevents antigen-stimulated exocytosis in mast cells without hindering Ca2 influx
Catalog No. 50-313-7796 Supplier Selleck Chemical LLC Supplier No. E1878100MG
Bromoenol lactone((6E)-Bromoenol lactone) is a selective irreversible potent inhibitor of calcium-independent phospholipase A2 (iPLA2 ) with an IC50 value of 7 M It prevents antigen-stimulated exocytosis in mast cells without hindering Ca2 influx