Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoethane 99.0+%, TCI America™
SDP

Catalog No. B058825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B058825G 25 g
B0588500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B058825G Supplier TCI America Supplier No. B058825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 74-96-4
Molecular Formula C2H5Br
Molecular Weight (g/mol) 108.966
MDL Number MFCD00000232
InChI Key RDHPKYGYEGBMSE-UHFFFAOYSA-N
Synonym ethyl bromide, ethane, bromo, 1-bromoethane, monobromoethane, bromic ether, hydrobromic ether, bromure d'ethyle, ethylbromide, etylu bromek, halon 2001
PubChem CID 6332
IUPAC Name bromoethane
SMILES CCBr

Specifications

Color Yellow
Boiling Point 38°C
Quantity 25 g
UN Number 1891
Formula Weight 108.97
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromoethane
EINECSNumber (9)-0518
RTECSNumber KH6475000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.