Learn More
Medchemexpress LLC Bromoethanol-d4 | 81764-55-8 | C2HD4BrO | 100 MG
SDP
Catalog No. 5000375192
Encompass_Preferred
Bromoethanol-d4 is the deuterium labeled Bromoethanol, intended for research use only. Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation during the drug development process. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Affects pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-75192 Supplier Medchemexpress LLC Supplier No. HY150535S100MG
Bromoethanol-d4 is the deuterium labeled Bromoethanol, intended for research use only. Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation during the drug development process. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Affects pharmacokinetic and metabolic profiles of drugs.