Learn More
Medchemexpress LLC Bromoethanol-d4 | 81764-55-8 | C2HD4BrO | 250 MG
SDP
Catalog No. 5000374538
Encompass_Preferred
Bromoethanol-d4 is the deuterium labeled Bromoethanol. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-74538 Supplier Medchemexpress LLC Supplier No. 250MGHY150535S
Bromoethanol-d4 is the deuterium labeled Bromoethanol. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs