Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromoform

Catalog No. 501861346
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-186-1346 50 g
50-186-1345 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-186-1346 Supplier Sigma Aldrich Supplier No. 24103250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bromoform
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-25-2
Density 2.89 g/mL (at 25°C (literature))
Molecular Formula CHBr3
Linear Formula CHBr3
MDL Number MFCD00000128
Melting Point 5°C to 8°C (lit.)
Boiling Point 146°C to 150°C (lit.)
Synonym Tribromomethane
Molecular Weight (g/mol) 252.73
Quantity 50 g
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.