Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoform, 96%, stab. with ethanol

Catalog No. AAA119040I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA119040I 5000 g
AAA1190430 250 g
AAA119040B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA119040I Supplier Thermo Scientific Chemicals Supplier No. A119040I
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Bromoform, 97%, CHBr3, CAS Number-75-25-2, tribromomethane, methenyl tribromide, methane, tribromo, tribrommethan, bromoforme, bromoformio, tribromometan, tribrommethaan, methyl tribromide, rcra waste number u225, 5000g, 1731048, 148 deg.C to 150 deg.C, CHEBI:38682, 2.81 | CAS: 75-25-2 | CHBr3 | 252.731 g/mol

Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.

Solubility
Slightly soluble in water.

Notes
Light-sensitive. A lachrymator. Incompatible with chemically active metals and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-25-2
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.731
MDL Number MFCD00000128
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br

Specifications

Melting Point 8°C to 9°C
Density 2.81
Boiling Point 148°C to 150°C
Flash Point None
Refractive Index 1.585
Quantity 5000 g
UN Number UN2515
Beilstein 1731048
Sensitivity Light sensitive
Merck Index 14,1420
Solubility Information Slightly soluble in water.
Formula Weight 252.73
Percent Purity 97%
Chemical Name or Material Bromoform, Stabilized with ethanol
Show More Show Less
Hazard Category H302-H315-H319-H331
Hazard Statement GHS H Statement
H351-H302-H332-H315-H319
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BROMOFORM
EINECSNumber 200-854-6
RTECSNumber PB5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.