Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoform amylene stabilized,, MilliporeSigma™ Supelco™

Catalog No. 111020418 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0418 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0418 Supplier MilliporeSigma Supelco Supplier No. 369721G
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-25-2
Vapor Pressure 5 mmHg (20°C)
Vapor Density 8.7 (vs. air)
Density 2.89 g/mL (at 25°C)
Physical Form Neat
Linear Formula CHBr3
MDL Number MFCD00000128
Melting Point 5°C to 8°C
Boiling Point 146°C to 150°C
Synonym Tribromomethane
Type Analytical Standard
Quantity 1 g
Formula Weight 252.73
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.