Learn More
Sigma Aldrich Fine Chemicals Biosciences Bromoform-d >=99.5 atom % D, >=99% (CP), contains copper as stabilizer | 2909-52-6 | MFCD00000129 | 25G
Catalog No. 501861342
Encompass
Bromoform-d >=99.5 atom % D, >=99% (CP), contains copper as stabilizer | Purity: >=99% (CP) | Mol Wt: 253.74 | 2909-52-6 | MFCD00000129 | 25G
Catalog No. 50-186-1342 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 32932025G
Bromoform-d >=99.5 atom % D, >=99% (CP), contains copper as stabilizer | Purity: >=99% (CP) | Mol Wt: 253.74 | 2909-52-6 | MFCD00000129 | 25G