Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoform (stabilized with 2-Methyl-2-butene) 99.0+%, TCI America™
SDP

Catalog No. B0806100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0806100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0806100ML Supplier TCI America Supplier No. B0806100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-25-2
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.731
MDL Number MFCD00000128
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br

Specifications

Color Colorless
Boiling Point 148°C
Quantity 100 mL
UN Number 2515
Formula Weight 252.73
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromoform (stabilized with 2-Methyl-2-butene) [for Spectrophotometry]
EINECSNumber (2)-0040
RTECSNumber PB5600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.