Learn More
AdipoGen Bromomethyl-7-methoxycoumarin
SDP
Supplier:  AdipoGen CDXB0022G001
Encompass_Preferred
Chemical. CAS 35231-44-8. Formula C11H9BrO3. MW 269.09. Synthetic. Blue fluorescent labeling reagent for fatty acids and carboxylates. Used for fluorometric analysis, derivatization, and TLC and HPLC separation of a range of naturally occurring acids. Used as a substrate for measuring histone acetyltransferase HATs, peptidases and sirtuin activity.
Catalog No. 50-256-1709