Learn More
AdipoGen Bromomethyl acetate
SDP
Supplier:  AdipoGen CDXB0074G005
Encompass_Preferred
Chemical. CAS 590-97-6. Formula C3H5BrO2. MW 152.97. Synthetic. Building block and reactant for synthesis. Used in the synthesis of optically active agents or in the conversion of ketones and aldehydes to 1,2-diacetates. Shown to have cytotoxic and mutagenic effects.
Catalog No. 50-256-1686