Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromomethyl acetate

Catalog No. 30320810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-320-810G 10 g
30-320-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-320-810G Supplier Sigma Aldrich Supplier No. 30320810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The interaction of bromomethyl acetate with o(6)-alkylguanine-DNA alkyltransferase (DNA repair protein) was studied in vitro.bromomethyl acetate was used in synthesis of optically active cyclohexene antisepsis agent, ethyl (6r)-6-[n-(2-chloro-4-fluorophenyl)sulfamoyl]cyclohex-1-ene-1-carboxylate [(r)-1: tak-242)]
  • It was also used as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates

Specifications

CAS 590-97-6
Density 1.56 g/mL (at 25°C (literature))
Boiling Point 130°C to 133°C (750 mmHg)
Molecular Formula C3H5BrO2
Linear Formula CH3CO2CH2Br
Refractive Index n20/D 1.447 (literature)
MDL Number MFCD00000170
Quantity 10 g
Molecular Weight (g/mol) 152.97
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.