Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Bromomethyl)chlorodimethylsilane 95.0+%, TCI America™
SDP

Catalog No. B084725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B084725G 25 g
B08475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B084725G Supplier TCI America Supplier No. B084725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16532-02-8
Molecular Formula C3H8BrClSi
Molecular Weight (g/mol) 187.536
MDL Number MFCD00000503
InChI Key CAURZYXCQQWBJO-UHFFFAOYSA-N
Synonym bromomethyl chlorodimethylsilane, bromomethyldimethylchlorosilane, bromomethyl dimethylchlorosilane, bromomethyl chloro dimethylsilane, bmdmcs, silane, bromomethyl chlorodimethyl, bromomethylchlorodimethylsilane, acmc-209dsc, bromomethyl-dimethyl-chlorosilane, caurzyxcqqwbjo-uhfffaoysa
PubChem CID 85476
IUPAC Name bromomethyl-chloro-dimethylsilane
SMILES C[Si](C)(CBr)Cl

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 g
UN Number 2986
Formula Weight 187.54
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (Bromomethyl)chlorodimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.