Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Bromomethyl)cyclohexane 97.0+%, TCI America™
SDP

Catalog No. B1708100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1708100G 100 g
B170825G 25 g
B1708500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1708100G Supplier TCI America Supplier No. B1708100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2550-36-9
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD00001509
InChI Key UUWSLBWDFJMSFP-UHFFFAOYSA-N
Synonym bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane
PubChem CID 137636
IUPAC Name (bromomethyl)cyclohexane
SMILES BrCC1CCCCC1

Specifications

Color Yellow
Boiling Point 94°C
Quantity 100 g
Formula Weight 177.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (Bromomethyl)cyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.