Learn More
Medchemexpress LLC (Bromomethyl)cyclopropane | 7051-34-5 | MFCD00001306 | 99.2% | 135.00 g/mol | C4H7Br | 25g
SDP
Supplier:  Medchemexpress LLC HYY110025G
Encompass_Preferred
Cyclopropylmethyl bromide is an intermediate Cyclopropylmethyl bromide can be used in the preparation of Firocoxib (HY-14670) Firocoxib (ML 1785713) is a potent selective orally active COX-2 inhibitor[1]
Catalog No. 50-002-68081