Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromomethyl methyl ether

Catalog No. 16175625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-175-625G 25 g
16-175-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-175-625G Supplier Sigma Aldrich Supplier No. 16175625G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Bromomethyl methyl ether was used in enantiocontrolled synthesis of intermediate containing carbons 18 to 35 of the macrocyclic immunosuppressant fk-506
  • It was used in the synthesis of 1-methoxymethyl-4-tritylthio-2-azetidinone

Specifications

CAS 13057-17-5
Density 1.531 g/mL (at 25°C (literature))
Boiling Point 87°C (lit.)
Molecular Formula C2H5BrO
Refractive Index n20/D 1.456 (literature)
Linear Formula BrCH2OCH3
MDL Number MFCD00000171
Quantity 25 g
Molecular Weight (g/mol) 124.96
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.