Learn More
Description
- Bromonitromethane has been used:
- In production of various protected α-bromo nitroalkane donors (including fmoc) for use in umpolung amide synthesis
- In preparation of (z)-1-bromo-1-nitroalkenes via sodium iodide-catalyzed henry reaction
- In diastereo- and enantioselective cyclopropanation of α,α-unsaturated a-ketoesters via domino michael-addition/intramolecular-alkylation strategy
Specifications
Specifications
| CAS | 563-70-2 |
| Density | 2.007 g/mL (at 25°C (literature)) |
| Boiling Point | 146°C to 148°C (750 mmHg) |
| Molecular Formula | CH2BrNO2 |
| Refractive Index | n20/D 1.496 (literature) |
| Linear Formula | BrCH2NO2 |
| MDL Number | MFCD00007401 |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 139.94 |
| Percent Purity | 90% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.