Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromonitromethane

Catalog No. 2558581G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-585-81G 1 g
25-585-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-585-81G Supplier Sigma Aldrich Supplier No. 2558581G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Bromonitromethane has been used:
  • In production of various protected α-bromo nitroalkane donors (including fmoc) for use in umpolung amide synthesis
  • In preparation of (z)-1-bromo-1-nitroalkenes via sodium iodide-catalyzed henry reaction
  • In diastereo- and enantioselective cyclopropanation of α,α-unsaturated a-ketoesters via domino michael-addition/intramolecular-alkylation strategy

Specifications

CAS 563-70-2
Density 2.007 g/mL (at 25°C (literature))
Boiling Point 146°C to 148°C (750 mmHg)
Molecular Formula CH2BrNO2
Refractive Index n20/D 1.496 (literature)
Linear Formula BrCH2NO2
MDL Number MFCD00007401
Quantity 1 g
Molecular Weight (g/mol) 139.94
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.