Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromopentafluorobenzene

Catalog No. B7515810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B7515810G 10 g
B7515850G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B7515810G Supplier Sigma Aldrich Supplier No. B7515810G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bromopentafluorobenzene

Specifications

CAS 344-04-7
Melting Point -31°C (lit.)
Density 1.981 g/mL (at 25°C (literature))
Boiling Point 137°C (lit.)
Molecular Formula C6BrF5
Refractive Index n20/D 1.449 (literature)
Linear Formula BrC6F5
MDL Number MFCD00000287
Quantity 10 g
Molecular Weight (g/mol) 246.96
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.