Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromopentafluorobenzene 99.0+%, TCI America™
SDP

Catalog No. B111625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B111625G 25 g
B11165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B111625G Supplier TCI America Supplier No. B111625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 344-04-7
Molecular Formula C6BrF5
Molecular Weight (g/mol) 246.96
MDL Number MFCD00000287
InChI Key XEKTVXADUPBFOA-UHFFFAOYSA-N
Synonym bromopentafluorobenzene, pentafluorobromobenzene, benzene, bromopentafluoro, bromoperfluorobenzene, pentafluorophenyl bromide, 2,3,4,5,6-pentafluorobromobenzene, benzene, 1-bromo-2,3,4,5,6-pentafluoro, 1-bromo-pentafluorobenzene, 2,3,4,5,6-bromopentafluorobenzene, 1-bromo-2,3,4,5,6-pentafluoro-benzene
PubChem CID 9578
IUPAC Name 1-bromo-2,3,4,5,6-pentafluorobenzene
SMILES FC1=C(F)C(F)=C(Br)C(F)=C1F

Specifications

Melting Point -31°C
Color Yellow
Boiling Point 137°C
Quantity 25 g
Formula Weight 246.96
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromopentafluorobenzene
EINECSNumber (3)-4285
RTECSNumber CY9040552
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.