Learn More
Bromophenol Blue, 0.1% Aqueous, Reagents
Specifications
Specifications
| Chemical Name or Material | Bromophenol Blue, 0.1% Aqueous |
| CAS | 7732-18-5, 62625-28-9 |
| Boiling Point | 100°C |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0001 |
| Color | Dark Reddish Purple |
| Synonym | 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Percent Purity | 0.1% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.