Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue, 0.1% w/v in denatured Ethanol, Reagents

Catalog No. 01335838
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-838 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-838 Supplier Reagents Holdings Llc Supplier No. CB004100100A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromophenol Blue, 0.1% w/v in denatured Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromophenol Blue, 0.1% w/v in denatured Ethanol
Boiling Point 72°C
CAS 64-17-5, 67-63-0, 67-63-0
Packaging Poly Bottle
Specific Gravity 0.8
Color Orange/Red
Synonym 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue
Quantity 100 mL
Physical Form Liquid
Percent Purity 0.1%
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.