Learn More
Bromophenol Blue, 0.5% (w/w) in IPA, Reagents
Specifications
Specifications
| Chemical Name or Material | Bromophenol Blue, 0.5% (w/w) in IPA |
| Boiling Point | 82°C |
| CAS | 67-63-0, 115-39-9 |
| Packaging | Poly Bottle |
| Specific Gravity | 0.781 |
| Synonym | 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue |
| Quantity | 1 L |
| Physical Form | Liquid |
| Percent Purity | 0.5% |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.