Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue, 1 g/L in 20% Ethanol, Reagents

Catalog No. 01335740
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-740 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-740 Supplier Reagents Holdings Llc Supplier No. CB1492001A
Add to Cart
Add to Cart

Bromophenol Blue, 1 g/L in 20% Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromophenol Blue, 1 g/L in 20% Ethanol
CAS 7732-18-5, 64-17-5, 64-17-5
Synonym 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue
Percent Purity 0.1%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.961
Quantity 1 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.