Learn More
Cayman Chemical Bromophenol Blue 100g
SDP
Catalog No. 502954218
Encompass_Preferred
Commonly used as a pH indicator, a color marker in agarose or polyacrylamide gel electrophoresis, and a dye to detect proteins and nucleic acids (absorbance at 610 nm), particularly when staining living tissues
Catalog No. 50-295-4218 Supplier Cayman Chemical Supplier No. 14331100 G
Commonly used as a pH indicator, a color marker in agarose or polyacrylamide gel electrophoresis, and a dye to detect proteins and nucleic acids (absorbance at 610 nm), particularly when staining living tissues