Learn More
Medchemexpress LLC Bromophenol blue | 115-39-9 | MFCD00005875 | 5 G
SDP
Supplier:  Medchemexpress LLC HYB15715G
Encompass_Preferred
Bromophenol blue is an acid phthalein dye utilized as a tracking dye for electrophoresis. It serves as a pH indicator with a transition range of pH 3 to 4.6. Moreover, it is used as a sensor for the determination of various compounds, including ammonia, drugs, proteins, and amino acids.
  • Functions as a pH indicator with a transition range of pH 3 to 4.6.
  • Serves as a sensor for determining compounds like ammonia, drugs, proteins, and amino acids.
  • Exhibits strong absorbance at 610 nm when bound to proteins.
Catalog No. 50-004-11725