Learn More
Cayman Chemical Bromophenol Blue 25g
SDP
Catalog No. 502952803
Encompass_Preferred
Commonly used as a pH indicator, a color marker in agarose or polyacrylamide gel electrophoresis, and a dye to detect proteins and nucleic acids (absorbance at 610 nm), particularly when staining living tissues
Catalog No. 50-295-2803 Supplier Cayman Chemical Supplier No. 1433125 G
Commonly used as a pH indicator, a color marker in agarose or polyacrylamide gel electrophoresis, and a dye to detect proteins and nucleic acids (absorbance at 610 nm), particularly when staining living tissues