Learn More
Sigma Aldrich Fine Chemicals Biosciences Bromophenol Blue, 115-39-9, MFCD00005875, 25 G
Catalog No. NC0768154
Encompass
Empirical Formula (Hill Notation): C19H10Br4O5S, Molecular Weight: 669.96, Synonym: 3′,3′′,5′,5′′-Tetrabromophenolsulfonephthalein, Bromophenol Blue (BPB), Bromphenol Blue Sultone Form. Bromophenol Blue has been used as a tracking dye for nucleic acid and protein electrophoresis in agarose and polyacrylamide gels.
Catalog No. NC0768154 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B012625G
Empirical Formula (Hill Notation): C19H10Br4O5S, Molecular Weight: 669.96, Synonym: 3′,3′′,5′,5′′-Tetrabromophenolsulfonephthalein, Bromophenol Blue (BPB), Bromphenol Blue Sultone Form. Bromophenol Blue has been used as a tracking dye for nucleic acid and protein electrophoresis in agarose and polyacrylamide gels.