Learn More
Bio Rad Laboratories BROMOPHENOL BLUE
Catalog No. NC9462352
Encompass
Bromophenol Blue is used as a tracking dye to monitor electrophoresis runs. Supplied as 10 g. (161-0404)
Catalog No. NC9462352 Supplier Bio Rad Laboratories Supplier No. 1610404
Bromophenol Blue is used as a tracking dye to monitor electrophoresis runs. Supplied as 10 g. (161-0404)