Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue, ACS Reagent (Sultone) not water soluble, Reagents

Catalog No. 01336775
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-775 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-775 Supplier Reagents Holdings Llc Supplier No. C61065005C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromophenol Blue, ACS Reagent (Sultone) not water soluble, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromophenol Blue, ACS Reagent (Sultone) not water soluble
CAS 115-39-9
Molecular Formula C19H10Br4O5S
Synonym 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue
Molecular Weight (g/mol) 669.9607
Grade ACS
Packaging Glass Bottle
Specific Gravity 0.954
Color Orange
Quantity 5 g
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.