Learn More
Bromophenol Blue, ACS Reagent (Sultone) not water soluble, Reagents
Specifications
Specifications
| Chemical Name or Material | Bromophenol Blue, ACS Reagent (Sultone) not water soluble |
| CAS | 115-39-9 |
| Molecular Formula | C19H10Br4O5S |
| Synonym | 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue |
| Molecular Weight (g/mol) | 669.9607 |
| Grade | ACS |
| Packaging | Glass Bottle |
| Specific Gravity | 0.954 |
| Color | Orange |
| Quantity | 1 g |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.