Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue, ACS, Sigma-Aldrich

Catalog No. 01258991 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-991 25 g
01-259-053 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-991 Supplier MilliporeSigma Supelco Supplier No. 11439125G
Add to Cart
Add to Cart

Bromophenol blue is present as a sulfonated hydroxyquinone in acidic aqueous solutions. It is also known as tetrabromophenolsulfonphthalein. Bromophenol Blue is a pH indicator and a dye appearing as a strong blue color. It has a slight negative charge and will migrate the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromophenol Blue
CAS 115-39-9
Molecular Weight (g/mol) 669.96 g/mol
Grade ACS
pH 3 to 4.6
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.