Learn More
Bromophenol Blue Indicator, 0.04% w/v, Reagents
Specifications
Specifications
| Chemical Name or Material | Bromophenol Blue Indicator, 0.04% w/v |
| CAS | 7732-18-5, 62625-28-9 |
| Boiling Point | 100°C |
| Synonym | 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue |
| Percent Purity | 0.04% |
| Grade | Reagent |
| Packaging | Glass Bottle |
| Specific Gravity | 1.001 |
| Color | Dark Reddish Purple |
| Quantity | 500 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.