Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue Indicator, 0.04% w/v, Reagents

Catalog No. 01336560
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass Bottle
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-560 500 mL Poly Bottle
01-336-563 100 mL Glass Bottle
01-336-562 1 L Glass Bottle
01-336-559 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-336-560 Supplier Reagents Holdings Llc Supplier No. C7106300500A
Only null left
Add to Cart
Add to Cart

Bromophenol Blue Indicator, 0.04% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromophenol Blue Indicator, 0.04% w/v
CAS 7732-18-5, 62625-28-9
Boiling Point 100°C
Synonym 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue
Percent Purity 0.04%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.001
Color Dark Reddish Purple
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.