Learn More
Crescent Chemical Co Inc BROMOPHENOL BLUE-NA-SALT
SDP
Supplier:  Crescent Chemical Co Inc SE15375.02
Encompass_Preferred
Bromophenol Blue-Na-salt Tracking dye for electrophoresis; indicator pH 3.4 (yellow) - 4.6 (purple). Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 15375.02 CAS 34725-61-6
Catalog No. 50-247-383