Learn More
Medchemexpress LLC Bromophenol blue sodium | 34725-61-6 | 691.94 g/mol | C19H9Br4NaO5S | 25 G
SDP
Catalog No. 5000209707
Encompass_Preferred
Bromophenol blue sodium is a biochemical reagent and pH indicator used as a tracking dye in nucleic acid and protein electrophoresis and as a reagent in colorimetric assays. It is supplied as a solid powder and is provided in a 25 g package.
  • Biochemical reagent and pH indicator.
  • Used as a tracking dye for nucleic acid and protein electrophoresis.
  • Supplied as a solid powder in a 25 g package.
  • Data sheet, SDS, and COA available for download.
  • Store at room temperature for solid; in solution store at -80°C (2 years) or -20°C (1 year).
Catalog No. 50-002-09707 Supplier Medchemexpress LLC Supplier No. 25GHYW088001
Bromophenol blue sodium is a biochemical reagent and pH indicator used as a tracking dye in nucleic acid and protein electrophoresis and as a reagent in colorimetric assays. It is supplied as a solid powder and is provided in a 25 g package.
  • Biochemical reagent and pH indicator.
  • Used as a tracking dye for nucleic acid and protein electrophoresis.
  • Supplied as a solid powder in a 25 g package.
  • Data sheet, SDS, and COA available for download.
  • Store at room temperature for solid; in solution store at -80°C (2 years) or -20°C (1 year).