Learn More
Bioworld Bromophenol Blue, Sodium Salt, 50 g
Catalog No. 50255653
Encompass_Preferred
An electrophoretic tracking dye for nucleic acid and protein gels.Used as a pH indicator for neutral or alkaline conditions.ACS Grade3',3'',5',5''-Tetrabromophenolsulphonephthalein Sodium SaltVisual transition interval pH 3.0-4.6Appearance: SolidMelting Point: >300 °CPH Value: 3.0-4.6
Catalog No. 50-255-653 Supplier Bioworld Supplier No. 402400162
An electrophoretic tracking dye for nucleic acid and protein gels.Used as a pH indicator for neutral or alkaline conditions.ACS Grade3',3'',5',5''-Tetrabromophenolsulphonephthalein Sodium SaltVisual transition interval pH 3.0-4.6Appearance: SolidMelting Point: >300 °CPH Value: 3.0-4.6