Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromophenol blue sodium salt | 34725-61-6 | MFCD00013793 | 691.94 | C19H9Br4NaO5S | 10 G
SDP

Catalog No. 5000208281
Encompass_Preferred

Bromophenol blue sodium salt is a sodium salt form of the pH indicator bromophenol blue supplied as a solid biochemical reagent for laboratory use. It is commonly used as a tracking dye and pH indicator in electrophoresis and buffer applications.

  • pH indicator and tracking dye for electrophoresis
  • Sodium salt solid form for buffer compatibility
  • Soluble in aqueous buffers used in nucleic acid and protein gels
  • Available in multiple pack sizes, including 10 g
  • Provided with datasheet, COA, and SDS on the manufacturer site

Catalog No. 50-002-08281 Supplier Medchemexpress LLC Supplier No. HYW08800110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromophenol blue sodium salt is a sodium salt form of the pH indicator bromophenol blue supplied as a solid biochemical reagent for laboratory use. It is commonly used as a tracking dye and pH indicator in electrophoresis and buffer applications.

  • pH indicator and tracking dye for electrophoresis
  • Sodium salt solid form for buffer compatibility
  • Soluble in aqueous buffers used in nucleic acid and protein gels
  • Available in multiple pack sizes, including 10 g
  • Provided with datasheet, COA, and SDS on the manufacturer site

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.