Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromophenol blue sodium salt | 34725-61-6 | MFCD00013793 | 691.94 g/mol | C19H9Br4NaO5S | 100 G
SDP

Catalog No. 5000208417
Encompass_Preferred

Bromophenol blue sodium salt is the sodium salt form of bromophenol blue, a pH indicator and electrophoretic tracking dye used in life-science research. It is supplied as a biochemical reagent for research use and is commonly used to monitor sample migration in nucleic acid and protein gel electrophoresis.

  • Used as an electrophoresis tracking dye for nucleic acid and protein gels.
  • Acts as a pH indicator, changing color around pH 3.0-4.6.
  • Available in multiple package sizes, including 100 g.
  • Molecular formula C19H9Br4NaO5S and molecular weight 691.94 g/mol.
  • CAS number 34725-61-6 for unambiguous identification.

Catalog No. 50-002-08417 Supplier Medchemexpress LLC Supplier No. HYW088001100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromophenol blue sodium salt is the sodium salt form of bromophenol blue, a pH indicator and electrophoretic tracking dye used in life-science research. It is supplied as a biochemical reagent for research use and is commonly used to monitor sample migration in nucleic acid and protein gel electrophoresis.

  • Used as an electrophoresis tracking dye for nucleic acid and protein gels.
  • Acts as a pH indicator, changing color around pH 3.0-4.6.
  • Available in multiple package sizes, including 100 g.
  • Molecular formula C19H9Br4NaO5S and molecular weight 691.94 g/mol.
  • CAS number 34725-61-6 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.