Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromophenol blue sodium salt | 34725-61-6 | MFCD00013793 | 691.94 g/mol | C19H9Br4NaO5S | 5 G
SDP

Catalog No. 5000208434
Encompass_Preferred

Bromophenol blue sodium salt is a water-soluble pH indicator and tracking dye used as a biochemical assay reagent. It is supplied as a brown to dark brown solid and is commonly used as a tracking dye in nucleic acid and protein electrophoresis, and as a pH indicator in buffer systems.

  • cas number: 34725-61-6
  • chemical formula: C19H9Br4NaO5S
  • molecular weight: 691.94 g/mol
  • appearance: solid, brown to dark brown
  • solubility: soluble in DMSO (100 mg/mL with ultrasonic warming)
  • applications: biochemical assay reagent, electrophoresis tracking dye
  • package sizes: includes 5 g, 10 g, 25 g, 50 g, 100 g

Catalog No. 50-002-08434 Supplier Medchemexpress LLC Supplier No. HYW0880015G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromophenol blue sodium salt is a water-soluble pH indicator and tracking dye used as a biochemical assay reagent. It is supplied as a brown to dark brown solid and is commonly used as a tracking dye in nucleic acid and protein electrophoresis, and as a pH indicator in buffer systems.

  • cas number: 34725-61-6
  • chemical formula: C19H9Br4NaO5S
  • molecular weight: 691.94 g/mol
  • appearance: solid, brown to dark brown
  • solubility: soluble in DMSO (100 mg/mL with ultrasonic warming)
  • applications: biochemical assay reagent, electrophoresis tracking dye
  • package sizes: includes 5 g, 10 g, 25 g, 50 g, 100 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.