Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS reagent

Catalog No. AC403160050
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC403160050 5 g
AC403160100 10 g
AC403160500 50 g
AC151351000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC403160050 Supplier Thermo Scientific Chemicals Supplier No. 403160050
Add to Cart

missing translation for 'validMainframeMsg'

Indicator | CAS: 34725-61-6 | C19H9Br4NaO5S | 691.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34725-61-6
Molecular Formula C19H9Br4NaO5S
Molecular Weight (g/mol) 691.94
MDL Number MFCD00013793
InChI Key KGANHHDZDYZJEM-UHFFFAOYNA-M
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein
PubChem CID 3791690
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate

Specifications

Melting Point >300.0°C
Packaging Glass bottle
Beilstein 19, I, 649
Quantity 5 g
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
Formula Weight 691.94
Grade ACS Reagent
Chemical Name or Material Bromophenol Blue, sodium salt, Pure
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.