Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue Sodium Salt, Fisher BioReagents

Catalog No. BP11425 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP114-25 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP114-25

missing translation for 'validMainframeMsg'

Bromophenol Blue Sodium Salt is commonly used as a tracking dye in electrophoresis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34725-61-6
Molecular Formula C19H9Br4NaO5S
Molecular Weight (g/mol) 691.94
InChI Key KGANHHDZDYZJEM-UHFFFAOYNA-M
Synonym 3,3',5,5'-Tetrabromophenolsulfonphthalein sodium salt
PubChem CID 3791690
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 300°C
Clarity Pass Test
Extinction Coefficient 800g(-1) cm(-1) min.
Identification Pass Test
Lambda Max 585 to 595nm
Formula Weight 691.94
Loss on Drying Run and Report %
Packaging Glass Bottle
Visual Transition Interval Pass Test
Color Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Bromophenol Blue Sodium Salt
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.