Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue Sodium Salt Indicator, Solstice

Catalog No. 60046167 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-167 50 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-167 Supplier Solstice Supplier No. 3276850G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34725-61-6
Molecular Formula C19H9Br4NaO5S
MDL Number MFCD00013793

Specifications

Packaging Glass Bottle
UN Number NONH for all modes of transport
Beilstein 6995701
Color Blue
Solubility Information Soluble in water
pH 4.6
Quantity 50 g
Physical Form Powder
Molecular Weight (g/mol) 691.95
Formula Weight 691.94g/mol
Grade Not Available
Chemical Name or Material Bromophenol Blue Sodium Salt
Show More Show Less
EINECSNumber 252-170-2
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.