Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Bromophenol Blue, Sodium Salt, Molecular biology grade, OmniPur™, Calbiochem™,

Catalog No. 283025GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-302-5GM 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-302-5GM Supplier MilliporeSigma™ Supplier No. 283025GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular biology grade dye used for tracking nucleic acids and proteins in electrophoresis gels or as a pH indicator.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34725-61-6
Molecular Formula C19H9Br4O5Sna
Molecular Weight (g/mol) 691.94
Synonym Tetrabromophenol Sulfonphthalein, Sodium Salt

Specifications

Packaging Glass Bottle
Color Purple to Red
Quantity 25 g
Physical Form Powder
Grade Molecular Biology
Chemical Name or Material Bromophenol Blue Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.