Learn More
Description
- Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.
- It is a tracking dye for alkaline and neutral buffer systems, as well as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.
Specifications
Specifications
| CAS | 115-39-9 |
| Molecular Formula | C19H10Br4O5S |
| Quantity | 10 g |
| InChI Key | UDSAIICHUKSCKT-UHFFFAOYSA-N |
| SMILES | C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br |
| IUPAC Name | 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
| Molecular Weight (g/mol) | 669.96 |
| PubChem CID | 8272 |
| Chemical Name or Material | Bromophenol Blue |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.