Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue Sodium Salt, Sigma-Aldrich

Catalog No. 01258992 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-992 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-992 Supplier MilliporeSigma Supelco Supplier No. 11440550G
Add to Cart
Add to Cart

Bromophenol Blue has been used in the sample buffer prepared for western blotting.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromophenol Blue Sodium Salt
CAS 34725-61-6
Molecular Weight (g/mol) 691.94 g/mol
Percent Purity 90%
Grade ACS
Color Blue
pH 3 to 4.6
Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.