Learn More
Abcam Bromophenol blue sodium salt, Tracking dye, 25G
Catalog No. 502704709
Encompass_Preferred
MW 691.94 g/mol. pH indicator (pH 3.0 (yellow) - 4.6 (blue)). Tracking dye for protein or nucleic acid electrophoresis in polyacrylamide or agarose gels.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4709 Supplier Abcam Supplier No. AB14633325G
MW 691.94 g/mol. pH indicator (pH 3.0 (yellow) - 4.6 (blue)). Tracking dye for protein or nucleic acid electrophoresis in polyacrylamide or agarose gels.
The product is subject to the following: Abcam Restricted Use Statement