Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue Solution, Sigma-Aldrich

Catalog No. 01258938 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-938 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-938 Supplier MilliporeSigma Supelco Supplier No. 318744500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tracking dye for nucleic acid or protein electrophoresis in agarose or polyacrylamide gels.

Bromophenol Blue is a tracking dye for nucleic acid or protein electrophoresis in agarose or polyacrylamide gels.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromophenol Blue
CAS 34725-61-6
Color Blue
pH 3 to 4.6
Quantity 500 mL
Molecular Weight (g/mol) 691.94 g/mol
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.