Learn More
Description
Bromophenol blue is present as a sulfonated hydroxyquinone in acidic aqueous solutions. It is also known as tetrabromophenolsulfonphthalein. Bromophenol Blue is a pH indicator and a dye appears a strong blue color. It has a slightly negative charge and will migrate in the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition.
Specifications
Specifications
| Chemical Name or Material | Bromophenol Blue |
| CAS | 115-39-9 |
| Color | Orange |
| pH | 3 to 4.6 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 669.96 g/mol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.