Learn More
Abcam Bromophenol blue, Tracking dye, 25G
Catalog No. 502704702
Encompass_Preferred
MW 669.96 g/mol. pH indicator (pH 3.0 (yellow) - 4.6 (blue)). Tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis. Migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4702 Supplier Abcam Supplier No. AB14633925G
MW 669.96 g/mol. pH indicator (pH 3.0 (yellow) - 4.6 (blue)). Tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis. Migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.
The product is subject to the following: Abcam Restricted Use Statement